Assay Details
| Gene Symbol: PLAAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076225.3 | NP_001069693.2 | 2 - 3 | 411 | 73 | |
| NM_001314065.1 | NP_001300994.1 | 2 - 3 | 110 | 73 | ||
| GenBank mRNA | BC114073.1 | - | 2 - 3 | 405 | 73 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC114073.1, NM_001076225.3, NM_001314065.1 transcripts.
Set Membership: |
|

PLAAT1
Genomic Map