Assay Details
| Gene Symbol: PLAAT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001076225.3 | NP_001069693.2 | 4 - 5 | 690 | 90 | |
| NM_001314065.1 | NP_001300994.1 | 4 - 5 | 389 | 90 | ||
| GenBank mRNA | BC114073.1 | - | 4 - 5 | 684 | 90 | |
More Information
Set Membership: |
|

PLAAT1
Genomic Map