Assay Details
| Gene Symbol: IRX4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001098466.1 | NP_001091936.1 | 3 - 4 | 575 | 77 | |
| GenBank mRNA | BC134434.1 | - | 3 - 4 | 575 | 77 | |
More Information
Set Membership: |
|

IRX4
Genomic Map