Assay Details
| Gene Symbol: PRMT9 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099051.1 | NP_001092521.1 | 8 - 9 | 1534 | 67 | |
| GenBank mRNA | BC142231.1 | - | 8 - 9 | 1534 | 67 | |
More Information
Set Membership: |
|

PRMT9
Genomic Map