Assay Details
| Gene Symbol: TMEM19 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101184.1 | NP_001094654.1 | 1 - 2 | 355 | 68 | |
| GenBank mRNA | BT030682.1 | - | 1 - 2 | 355 | 68 | |
More Information
Set Membership: |
|

TMEM19
Genomic Map