Assay Details
| Gene Symbol: GMFB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101237.1 | NP_001094707.1 | 6 - 7 | 430 | 107 | |
| GenBank mRNA | BC151658.1 | - | 6 - 7 | 430 | 107 | |
| EH141890.1 | - | 6 - 7 | 430 | 107 | ||
More Information
Set Membership: |
|

GMFB
Genomic Map