Assay Details
| Gene Symbol: GABRG1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001101250.1 | NP_001094720.1 | 1 - 2 | 170 | 91 | |
| GenBank mRNA | BC149937.1 | - | 1 - 2 | 170 | 91 | |
More Information
Set Membership: |
|

GABRG1
Genomic Map