Assay Details
| Gene Symbol: POC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102158.1 | NP_001095628.1 | 11 - 12 | 1716 | 77 | |
| GenBank mRNA | BC149510.1 | - | 11 - 12 | 1716 | 77 | |
More Information
Set Membership: |
|

POC5
Genomic Map