Assay Details
| Gene Symbol: YDJC | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102221.1 | NP_001095691.1 | 1 - 2 | 202 | 68 | |
| GenBank mRNA | BC149866.1 | - | 1 - 2 | 202 | 68 | |
More Information
Set Membership: |
|

YDJC
Genomic Map