Assay Details
| Gene Symbol: NAGLU | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102226.2 | NP_001095696.1 | 5 - 6 | 1056 | 87 | |
| GenBank mRNA | BC148147.1 | - | 5 - 6 | 1057 | 87 | |
More Information
Set Membership: |
|

NAGLU
Genomic Map