Assay Details
Gene Symbol: KRT40 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001105418.1 | NP_001098888.1 | 2 - 3 | 582 | 63 | |
GenBank mRNA | BC134653.1 | - | 2 - 3 | 582 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
