Assay Details
| Gene Symbol: LRATD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105632.3 | NP_001099102.2 | 1 - 2 | 189 | 132 | |
| GenBank mRNA | BC153243.1 | - | 1 - 2 | 190 | 132 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC153243.1, NM_001105632.3 transcripts.
Set Membership: |
|

LRATD2
Genomic Map