Assay Details
| Gene Symbol: NUPR1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114515.2 | NP_001107987.1 | 2 - 3 | 283 | 75 | |
| GenBank mRNA | BC103251.1 | - | 2 - 3 | 285 | 75 | |
More Information
Set Membership: |
|

NUPR1
Genomic Map