Assay Details
| Gene Symbol: ROMO1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001114528.2 | NP_001108000.1 | 1 - 2 | 85 | 83 | |
| GenBank mRNA | BC102685.1 | - | 1 - 2 | 86 | 83 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC102685.1, NM_001114528.2 transcripts.
Set Membership: |
|

ROMO1
Genomic Map