Assay Details
Gene Symbol: ROMO1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001114528.2 | NP_001108000.1 | 2 - 3 | 217 | 88 | |
GenBank mRNA | BC102685.1 | - | 2 - 3 | 218 | 88 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() ![]() |
Gene Ontology Categories:
