Assay Details
| Gene Symbol: ZFP30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102539.1 | NP_001096009.1 | 1 - 2 | 85 | 88 | |
| GenBank mRNA | BC151289.1 | - | 1 - 2 | 85 | 88 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC151289.1, NM_001102539.1 transcripts.
Set Membership: |
|

ZFP30
Genomic Map