Assay Details
| Gene Symbol: ZFP30 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001102539.1 | NP_001096009.1 | 6 - 7 | 635 | 151 | |
| GenBank mRNA | BC151289.1 | - | 6 - 7 | 635 | 151 | |
More Information
Set Membership: |
|

ZFP30
Genomic Map