Assay Details
| Gene Symbol: ZNF613 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103337.1 | NP_001096807.1 | 3 - 4 | 325 | 84 | |
| GenBank mRNA | BC134644.1 | - | 3 - 4 | 325 | 84 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC134644.1, NM_001103337.1 transcripts.
Set Membership: |
|

ZNF613
Genomic Map