Assay Details
| Gene Symbol: ZNF613 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001103337.1 | NP_001096807.1 | 5 - 6 | 539 | 95 | |
| GenBank mRNA | BC134644.1 | - | 5 - 6 | 539 | 95 | |
More Information
Set Membership: |
|

ZNF613
Genomic Map