Assay Details
| Gene Symbol: ZNF771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113729.2 | NP_001107201.1 | 1 - 2 | 83 | 170 | |
| GenBank mRNA | BC151714.1 | - | 1 - 2 | 84 | 170 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC151714.1, NM_001113729.2 transcripts.
Set Membership: |
|

ZNF771
Genomic Map