Assay Details
| Gene Symbol: ZNF771 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001113729.2 | NP_001107201.1 | 2 - 3 | 236 | 118 | |
| GenBank mRNA | BC151714.1 | - | 2 - 3 | 237 | 118 | |
More Information
Set Membership: |
|

ZNF771
Genomic Map