Assay Details
| Gene Symbol: RNF186 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001035311.2 | NP_001030388.1 | 1 | 645 | 161 | |
| GenBank mRNA | BC102202.1 | - | 1 | 646 | 161 | |
More Information
Set Membership: |
|

RNF186
Genomic Map