Assay Details
| Gene Symbol: GTPBP8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001075828.2 | NP_001069296.1 | 1 | 251 | 138 | |
| GenBank mRNA | BC120144.1 | - | 1 | 252 | 138 | |
More Information
Set Membership: |
|

GTPBP8
Genomic Map