Assay Details
| Gene Symbol: TTC30B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099091.1 | NP_001092561.1 | 1 | 656 | 88 | |
| GenBank mRNA | BC146102.1 | - | 1 | 656 | 88 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TTC30B| Gene Symbol: TTC30B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099091.1 | NP_001092561.1 | 1 | 656 | 88 | |
| GenBank mRNA | BC146102.1 | - | 1 | 656 | 88 | |
Set Membership: |
|