Assay Details
| Gene Symbol: ZNF444 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001105649.1 | NP_001099119.1 | 1 | 43 | 138 | |
| GenBank mRNA | BC153298.1 | - | 1 | 43 | 138 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC153298.1, NM_001105649.1 transcripts.
Set Membership: |
|

ZNF444
Genomic Map