Assay Details
| Gene Symbol: HSPA1A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_203322.3 | NP_976067.3 | 1 | 85 | 145 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_203322.3 transcripts.
Set Membership: |
|

HSPA1A
Genomic Map