Assay Details
Gene Symbol: PRPS1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001046189.2 | NP_001039654.1 | 7 | 1687 | 162 | |
GenBank mRNA | BC113300.1 | - | 7 | 1648 | 162 | |
BC142427.1 | - | 7 | 1688 | 162 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
