Assay Details
| Gene Symbol: PRPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046189.2 | NP_001039654.1 | 2 - 3 | 456 | 74 | |
| GenBank mRNA | BC113300.1 | - | 2 - 3 | 417 | 74 | |
| BC142427.1 | - | 2 - 3 | 457 | 74 | ||
More Information
Set Membership: |
|

PRPS1
Genomic Map