Assay Details
| Gene Symbol: PRPS1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001046189.2 | NP_001039654.1 | 4 - 5 | 685 | 85 | |
| GenBank mRNA | BC113300.1 | - | 4 - 5 | 646 | 85 | |
| BC142427.1 | - | 4 - 5 | 686 | 85 | ||
More Information
Set Membership: |
|

PRPS1
Genomic Map