Assay Details
| Gene Symbol: NGF | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099362.1 | NP_001092832.1 | 1 | 649 | 75 | |
| GenBank mRNA | BC146037.1 | - | 1 | 649 | 75 | |
| M26809.1 | - | 1 | 293 | 75 | ||
More Information
Set Membership: |
|

NGF
Genomic Map