Assay Details
| Gene Symbol: UBE2I | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099372.1 | NP_001092842.1 | 2 - 3 | 162 | 71 | |
| GenBank mRNA | BC146107.1 | - | 2 - 3 | 162 | 71 | |
More Information
Set Membership: |
|

UBE2I
Genomic Map