Assay Details
| Gene Symbol: CCDC59 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001099390.1 | NP_001092860.1 | 2 - 3 | 855 | 100 | |
| GenBank mRNA | BC142178.1 | - | 2 - 3 | 855 | 100 | |
More Information
Set Membership: |
|

CCDC59
Genomic Map