Assay Details
| Gene Symbol: SPESP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001079773.1 | NP_001073241.1 | 1 - 2 | 140 | 66 | |
| GenBank mRNA | BC110032.1 | - | 1 - 2 | 140 | 66 | |
More Information
Set Membership: |
|

SPESP1
Genomic Map