Assay Details
| Gene Symbol: FAM83B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205795.1 | NP_001192724.1 | 1 - 2 | 66 | 94 | |
| GenBank mRNA | BC151752.1 | - | 1 - 2 | 66 | 94 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the BC151752.1, NM_001205795.1 transcripts.
Set Membership: |
|

FAM83B
Genomic Map