Assay Details
| Gene Symbol: FAM83B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205795.1 | NP_001192724.1 | 4 - 5 | 862 | 75 | |
| GenBank mRNA | BC151752.1 | - | 4 - 5 | 862 | 75 | |
More Information
Set Membership: |
|

FAM83B
Genomic Map