Assay Details
| Gene Symbol: TMPRSS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205892.1 | NP_001192821.1 | 10 - 11 | 1286 | 61 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

TMPRSS7| Gene Symbol: TMPRSS7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205892.1 | NP_001192821.1 | 10 - 11 | 1286 | 61 | |
Set Membership: |
|