Assay Details
| Gene Symbol: HS3ST2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205994.1 | NP_001192923.1 | 1 - 2 | 629 | 94 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

HS3ST2| Gene Symbol: HS3ST2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001205994.1 | NP_001192923.1 | 1 - 2 | 629 | 94 | |
Set Membership: |
|