Assay Details
| Gene Symbol: MIEF1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206126.1 | NP_001193055.1 | 1 - 2 | 147 | 91 | |
| GenBank mRNA | BC148943.1 | - | 3 - 4 | 547 | 91 | |
More Information
Set Membership: |
|

MIEF1
Genomic Map