Assay Details
| Gene Symbol: SYDE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192598.1 | NP_001179527.1 | 5 - 6 | 2832 | 102 | |
More Information
Set Membership: |
|
Please enter the information below and press OK to send your cart to Core Services for purchase.

SYDE2| Gene Symbol: SYDE2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192598.1 | NP_001179527.1 | 5 - 6 | 2832 | 102 | |
Set Membership: |
|