Assay Details
| Gene Symbol: AHDC1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206409.1 | NP_001193338.1 | 3 - 4 | 365 | 128 | |
More Information
Additional Information:
This assay is in the 5'UTR region of the NM_001206409.1 transcripts.
Set Membership: |
|

AHDC1
Genomic Map