Assay Details
| Gene Symbol: SIT1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206418.1 | NP_001193347.1 | 3 - 4 | 362 | 60 | |
| GenBank mRNA | BC149731.1 | - | 3 - 4 | 363 | 60 | |
More Information
Set Membership: |
|

SIT1
Genomic Map