Assay Details
Gene Symbol: PCBD1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206474.1 | NP_001193403.1 | 2 - 3 | 251 | 69 | |
GenBank mRNA | BC103434.1 | - | 2 - 3 | 253 | 69 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() ![]() |
Panther Classification:
Gene Ontology Categories:
