Assay Details
| Gene Symbol: ETV5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192834.2 | NP_001179763.1 | 11 - 12 | 1434 | 69 | |
| GenBank mRNA | BC151692.1 | - | 11 - 12 | 1435 | 69 | |
More Information
Set Membership: |
|

ETV5
Genomic Map