Assay Details
| Gene Symbol: SLC27A2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192863.1 | NP_001179792.1 | 1 - 2 | 481 | 65 | |
| GenBank mRNA | KP735930.1 | - | 1 - 2 | 412 | 65 | |
More Information
Set Membership: |
|

SLC27A2
Genomic Map