Assay Details
| Gene Symbol: ST8SIA5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001192874.2 | NP_001179803.1 | 6 - 7 | 1046 | 56 | |
| GenBank mRNA | AJ868432.1 | - | 6 - 7 | 663 | 56 | |
More Information
Set Membership: |
|

ST8SIA5
Genomic Map