Assay Details
| Gene Symbol: SREK1IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206619.1 | NP_001193548.1 | 1 - 2 | 190 | 97 | |
| GenBank mRNA | BC118474.1 | - | 1 - 2 | 135 | 97 | |
| EV629429.1 | - | 1 - 2 | 195 | 97 | ||
More Information
Additional Information:
This assay is in the 5'UTR region of the BC118474.1, NM_001206619.1 transcripts.
Set Membership: |
|

SREK1IP1
Genomic Map