Assay Details
| Gene Symbol: TBC1D12 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206699.2 | NP_001193628.2 | 3 - 4 | 1293 | 71 | |
| GenBank mRNA | EH376280.1 | - | 3 - 4 | 303 | 71 | |
More Information
Set Membership: |
|

TBC1D12
Genomic Map