Assay Details
Gene Symbol: MANEA | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | NM_001206725.1 | NP_001193654.1 | 1 - 2 | 104 | 94 | |
GenBank mRNA | BC103138.1 | - | 1 - 2 | 106 | 94 |

More Information
Additional Information:
This assay is in the 5'UTR region of the BC103138.1, NM_001206725.1 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
