Assay Details
| Gene Symbol: MANEA | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001206725.1 | NP_001193654.1 | 2 - 3 | 687 | 93 | |
| GenBank mRNA | BC103138.1 | - | 2 - 3 | 689 | 93 | |
| JN385276.1 | - | 1 - 2 | 571 | 93 | ||
| JQ946519.1 | - | 1 - 2 | 544 | 93 | ||
| JQ946520.1 | - | 1 - 2 | 544 | 93 | ||
| JQ946521.1 | - | 1 - 2 | 544 | 93 | ||
| JQ946522.1 | - | 1 - 2 | 544 | 93 | ||
More Information
Set Membership: |
|

MANEA
Genomic Map