Assay Details
| Gene Symbol: TRPC5 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
| RefSeq | NM_001166574.1 | NP_001160046.1 | 5 - 6 | 1703 | 63 | |
| GenBank mRNA | AJ271070.1 | - | 1 - 2 | 140 | 63 | |
More Information
Set Membership: |
|

TRPC5
Genomic Map